Product Details
- SNP ID
-
hCV30373893
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:123977765 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCATAGTGAGCTTAAACTTTTT[C/T]TAAGCACACACATACTTAGGAAGTG
- Phenotype
-
MIM: 300826
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STAG2
PubMed Links
Gene Details
- Gene
- STAG2
- Gene Name
- stromal antigen 2
There are no transcripts associated with this gene.
View Full Product Details