Product Details
- SNP ID
-
rs6070006
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:57170959 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAAGGGTCTGAATTCTCGGAGGAG[A/C]TAGTGGCAGCCACAGGCCCGGACCA
- Phenotype
-
MIM: 112267
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BMP7
PubMed Links
Gene Details
- Gene
- BMP7
- Gene Name
- bone morphogenetic protein 7
There are no transcripts associated with this gene.
View Full Product Details