Product Details

SNP ID
rs6001393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39031237 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGAAACTCAATGAATAAAACGCC[C/G]TGGGGCCTACTATGCACCCAGAAAA
Phenotype
MIM: 609900 MIM: 608993
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOBEC3D PubMed Links

Gene Details

Gene
APOBEC3D
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152426.3 Intron NP_689639.2
XM_005261337.3 Intron XP_005261394.1
XM_017028596.1 Intron XP_016884085.1
Gene
APOBEC3F
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006666.1 Intron NP_001006667.1
NM_145298.5 Intron NP_660341.2
XM_011529994.2 Intron XP_011528296.1
XM_017028642.1 Intron XP_016884131.1
XM_017028643.1 Intron XP_016884132.1

View Full Product Details