Product Details

SNP ID
rs10202823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171923622 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAGGGCATTCCTCATAGTCTCTC[C/T]ACATTAACCATTCCATTTTTATGAA
Phenotype
MIM: 603053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAT1 PubMed Links

Gene Details

Gene
HAT1
Gene Name
histone acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003642.3 758 Intron NP_003633.1
XM_006712808.3 758 Intron XP_006712871.1
XM_011512027.2 758 Intron XP_011510329.1
XM_017005136.1 758 UTR 5 XP_016860625.1
XM_017005137.1 758 UTR 5 XP_016860626.1

View Full Product Details