Product Details
- SNP ID
-
rs10513565
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:161498201 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATCTGGCTGGTAAGCTGCTTGGC[A/G]GTGAAAATCCCAAGGGCATGGTCAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OTOL1
PubMed Links
Gene Details
- Gene
- OTOL1
- Gene Name
- otolin 1
View Full Product Details