Product Details

SNP ID
rs10085741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1547021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTTCCTGCTCCCGCCGGAGCC[A/G]CCGCTGGGGTGGGTGCCGGGCCTGG
Phenotype
MIM: 600197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFK PubMed Links

Gene Details

Gene
MAFK
Gene Name
MAF bZIP transcription factor K
There are no transcripts associated with this gene.

Gene
TMEM184A
Gene Name
transmembrane protein 184A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097620.1 2482 Silent Mutation GGC,GGT G391G NP_001091089.1
XM_006715653.1 2482 Missense Mutation CGG,TGG R411W XP_006715716.1
XM_006715655.2 2482 Silent Mutation GGC,GGT G370G XP_006715718.1
XM_011515164.1 2482 Missense Mutation CGG,TGG R391W XP_011513466.1
XM_011515165.2 2482 Intron XP_011513467.1
XM_011515166.2 2482 Intron XP_011513468.1
XM_017011815.1 2482 UTR 5 XP_016867304.1

View Full Product Details