Product Details

SNP ID
rs9377663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:104684828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCTAACATGTATTTATTTTAAG[A/C]TCAACATAGCTTTATTTTGGATCTA
Phenotype
MIM: 610876
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HACE1 PubMed Links

Gene Details

Gene
HACE1
Gene Name
HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321080.1 Intron NP_001308009.1
NM_001321083.1 Intron NP_001308012.1
NM_001321084.1 Intron NP_001308013.1
NM_020771.3 Intron NP_065822.2
XM_017011113.1 Intron XP_016866602.1
XM_017011114.1 Intron XP_016866603.1
XM_017011115.1 Intron XP_016866604.1
XM_017011116.1 Intron XP_016866605.1
XM_017011118.1 Intron XP_016866607.1
XM_017011119.1 Intron XP_016866608.1
XM_017011120.1 Intron XP_016866609.1
XM_017011121.1 Intron XP_016866610.1
XM_017011122.1 Intron XP_016866611.1

View Full Product Details