Product Details
- SNP ID
-
rs9427942
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
24
- Location
-
Chr.1:196977318 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAAAAAAAAAAACCCAGGGGGCC[A/G]GCGGTGGGGAGCTTCCTCTCTCCTC
- Phenotype
-
MIM: 608593
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CFHR5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115846544] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CFHR5
- Gene Name
- complement factor H related 5
View Full Product Details