Product Details

SNP ID
rs10419534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49521011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGATCCACCCTCCCTGAAAGGAAC[C/T]TGGGCAGTAATGCCCAGTCCTAGGC
Phenotype
MIM: 601437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCGRT PubMed Links

Gene Details

Gene
FCGRT
Gene Name
Fc fragment of IgG receptor and transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136019.2 Intron NP_001129491.1
NM_004107.4 Intron NP_004098.1
XM_005258657.1 Intron XP_005258714.1
XM_011526627.2 Intron XP_011524929.1
Gene
RCN3
Gene Name
reticulocalbin 3
There are no transcripts associated with this gene.

View Full Product Details