Product Details

SNP ID
rs4540386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:108450562 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATACCGTAGTTAAGACTTAATGT[A/G]TGGCTTATATGGAATGCTTTCAATA
Phenotype
MIM: 607722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC2 PubMed Links

Gene Details

Gene
EMC2
Gene Name
ER membrane protein complex subunit 2
There are no transcripts associated with this gene.

View Full Product Details