Product Details
- SNP ID
-
rs9484199
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:138696520 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAACTTTTTTATTCTCTCAGGTAA[C/T]AGAAAATCCTGTTACTTTAAGAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ46906
PubMed Links
Gene Details
- Gene
- FLJ46906
- Gene Name
- uncharacterized LOC441172
There are no transcripts associated with this gene.
- Gene
- NHSL1
- Gene Name
- NHS like 1
There are no transcripts associated with this gene.
View Full Product Details