Product Details
- SNP ID
-
rs7083877
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:43467994 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGTCCAATTAACAACATTAATAG[A/G]AATTTGGAAGAACTTGAATCCAACC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF487
PubMed Links
Gene Details
- Gene
- ZNF487
- Gene Name
- zinc finger protein 487
There are no transcripts associated with this gene.
View Full Product Details