Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001287754.1 | 279 | Intron | NP_001274683.1 | ||
NM_001320143.1 | 279 | UTR 5 | NP_001307072.1 | ||
NM_001320144.1 | 279 | Intron | NP_001307073.1 | ||
NM_025228.3 | 279 | Intron | NP_079504.2 | ||
XM_005273279.4 | 279 | Intron | XP_005273336.1 | ||
XM_005273280.2 | 279 | Intron | XP_005273337.1 | ||
XM_011510018.2 | 279 | Intron | XP_011508320.1 | ||
XM_011510019.1 | 279 | Intron | XP_011508321.1 | ||
XM_017002399.1 | 279 | Intron | XP_016857888.1 | ||
XM_017002400.1 | 279 | Intron | XP_016857889.1 |