Product Details

SNP ID
rs10177500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108292508 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAATAAGCATGAGAAGATGCTCAA[C/T]ATCATTAGACATTAGGGGAATGCAA
Phenotype
MIM: 602385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SULT1C2 PubMed Links

Gene Details

Gene
SULT1C2
Gene Name
sulfotransferase family 1C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001056.3 Intron NP_001047.1
NM_176825.2 Intron NP_789795.1
XM_005264013.3 Intron XP_005264070.1

View Full Product Details