Product Details

SNP ID
rs9312181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67470861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGAGTATCTGTAGTTGCTAACC[A/T]AAGGTTACAGTCCTAGTTCTTGAAG
Phenotype
MIM: 117141
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CENPC PubMed Links

Gene Details

Gene
CENPC
Gene Name
centromere protein C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001812.2 4798 Intron NP_001803.2
XM_006714064.2 4798 UTR 3 XP_006714127.1
XM_011531542.2 4798 Intron XP_011529844.1

View Full Product Details