Product Details
- SNP ID
-
hCV30414056
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112490905 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATATTTTAATACAACCTAACTGAA[A/G]TCCTATCTGTCATGATAGAAAGAAA
- Phenotype
-
MIM: 608291
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTL
PubMed Links
Gene Details
- Gene
- TTL
- Gene Name
- tubulin tyrosine ligase
There are no transcripts associated with this gene.
View Full Product Details