Product Details

SNP ID
rs8179500
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:211337141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGACAGAGAGCAAAGACATGAA[C/T]AAGTAATATGTTAGGCAGTAATAAG
Phenotype
MIM: 602356
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAF5 PubMed Links

Gene Details

Gene
TRAF5
Gene Name
TNF receptor associated factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033910.2 Intron NP_001029082.1
NM_001319207.1 Intron NP_001306136.1
NM_004619.3 Intron NP_004610.1
NM_145759.2 Intron NP_665702.1
XM_011509957.2 Intron XP_011508259.1
XM_011509959.2 Intron XP_011508261.1
XM_011509960.2 Intron XP_011508262.1
XM_017002221.1 Intron XP_016857710.1

View Full Product Details