Product Details

SNP ID
rs10495746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24080010 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTTAGGAAGTGTCCGCACTAGGT[A/T]ATTGGAATCATAGGCTTTCTAGCTT
Phenotype
MIM: 607835 MIM: 605171
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM228B PubMed Links

Gene Details

Gene
FAM228B
Gene Name
family with sequence similarity 228 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145710.1 Intron NP_001139182.1
NM_001291328.1 Intron NP_001278257.1
Gene
SF3B6
Gene Name
splicing factor 3b subunit 6
There are no transcripts associated with this gene.

Gene
TP53I3
Gene Name
tumor protein p53 inducible protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206802.2 Intron NP_001193731.1
NM_004881.4 Intron NP_004872.2
NM_147184.3 Intron NP_671713.1
XM_005264650.2 Intron XP_005264707.1
XM_006712150.2 Intron XP_006712213.1

View Full Product Details