Product Details

SNP ID
rs10026631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82425953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTAGTAAAAACAGGCTACATAG[C/T]ATTTTGTTTTTACGTTTCATTTGTC
Phenotype
MIM: 607137
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENOPH1 PubMed Links

Gene Details

Gene
ENOPH1
Gene Name
enolase-phosphatase 1
There are no transcripts associated with this gene.

Gene
HNRNPDL
Gene Name
heterogeneous nuclear ribonucleoprotein D like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207000.1 Intron NP_001193929.1
NM_031372.3 Intron NP_112740.1

View Full Product Details