Product Details

SNP ID
rs5979375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:11141541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGAAACACTGAAAAATACTGGTA[A/G]GAAAGAAATGCAAATCAAATTAACA
Phenotype
MIM: 300118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP6 PubMed Links

Gene Details

Gene
ARHGAP6
Gene Name
Rho GTPase activating protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287242.1 Intron NP_001274171.1
NM_006125.2 Intron NP_006116.2
NM_013423.2 Intron NP_038267.1
NM_013427.2 Intron NP_038286.2

View Full Product Details