Product Details

SNP ID
rs5959656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:80022227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCTCCCTCCCTAACCCAGTTCA[A/G]GTTGAAACTTTGCTGCAAAGAATCC
Phenotype
MIM: 300307
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBX22 PubMed Links

Gene Details

Gene
TBX22
Gene Name
T-box 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109878.1 Intron NP_001103348.1
NM_001109879.1 Intron NP_001103349.1
NM_001303475.1 Intron NP_001290404.1
NM_016954.2 Intron NP_058650.1

View Full Product Details