Product Details

SNP ID
rs10404355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12169521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACTTTTGAATGTGGGTAAAGGTA[A/G]ACAAATAAGCCTTGGGCATTTCTGG
Phenotype
MIM: 604078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF136 PubMed Links

Gene Details

Gene
ZNF136
Gene Name
zinc finger protein 136
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003437.3 Intron NP_003428.1
XM_011528269.2 Intron XP_011526571.1
XM_017027243.1 Intron XP_016882732.1

View Full Product Details