Product Details

SNP ID
rs9626069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43962590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTTTAATGTGAATGAGACTTT[C/T]TGAAATGAAAAAACCTGTCAGGGTC
Phenotype
MIM: 612058
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAMM50 PubMed Links

Gene Details

Gene
SAMM50
Gene Name
SAMM50 sorting and assembly machinery component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015380.4 Intron NP_056195.3

View Full Product Details