Product Details

SNP ID
rs5763760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30089335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATTAAGACTTTGTAAAGAAAA[A/G]GGAAAATTTGCTGCTGCTTCTTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HORMAD2 PubMed Links

Gene Details

Gene
HORMAD2
Gene Name
HORMA domain containing 2
There are no transcripts associated with this gene.

Gene
HORMAD2-AS1
Gene Name
HORMAD2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details