Product Details
- SNP ID
-
rs10107827
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:23103142 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATACCATGGCCCGGATCCCCAAGAC[C/T]CTAAAGTTCGTCGTCGTCATCGTCG
- Phenotype
-
MIM: 603613
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC254896
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10107826] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC254896
- Gene Name
- uncharacterized LOC254896
There are no transcripts associated with this gene.
- Gene
- TNFRSF10C
- Gene Name
- TNF receptor superfamily member 10c
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003841.3 |
329 |
Silent Mutation |
ACC,ACT |
T7T |
NP_003832.2 |
View Full Product Details