Product Details

SNP ID
rs7255053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48005384 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCAGAGGCAGGAAAAGCGGATAG[A/C]TTCAGGAGCTCCAGAAAGTAGAACC
Phenotype
MIM: 607443
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELSPBP1 PubMed Links

Gene Details

Gene
ELSPBP1
Gene Name
epididymal sperm binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022142.4 Intron NP_071425.3
XM_017027130.1 Intron XP_016882619.1

View Full Product Details