Product Details

SNP ID
rs6117897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:834279 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATCACATATCCCCGCGCCCCACA[C/T]TGTCCTCGGGATACCCCCTTTGGGC
Phenotype
MIM: 611393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM110A PubMed Links

Gene Details

Gene
FAM110A
Gene Name
family with sequence similarity 110 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042353.2 Intron NP_001035812.1
NM_001289145.1 Intron NP_001276074.1
NM_001289146.1 Intron NP_001276075.1
NM_001289147.1 Intron NP_001276076.1
NM_031424.5 Intron NP_113612.1
NM_207121.4 Intron NP_997004.1
XM_011529381.2 Intron XP_011527683.1

View Full Product Details