Product Details

SNP ID
rs9311331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42937990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTTTTGAATATCTCTGATCCA[A/T]CTGTTGGCAGCTCTTTGTATTTCAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRBOX1 PubMed Links

Gene Details

Gene
KRBOX1
Gene Name
KRAB box domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205272.1 Intron NP_001192201.1
Gene
KRBOX1-AS1
Gene Name
KRBOX1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details