Product Details

SNP ID
rs6642839
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:110945936 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCAAACAGCAGACATTTTTTGCAT[A/G]CCTGCTATGTGTTATTAATGTTAAG
Phenotype
MIM: 300142
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PAK3 PubMed Links

Gene Details

Gene
PAK3
Gene Name
p21 (RAC1) activated kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128166.2 Intron NP_001121638.1
NM_001128167.2 Intron NP_001121639.1
NM_001128168.2 Intron NP_001121640.1
NM_001128172.2 Intron NP_001121644.1
NM_001128173.2 Intron NP_001121645.1
NM_001324325.1 Intron NP_001311254.1
NM_001324326.1 Intron NP_001311255.1
NM_001324327.1 Intron NP_001311256.1
NM_001324328.1 Intron NP_001311257.1
NM_001324329.1 Intron NP_001311258.1
NM_001324330.1 Intron NP_001311259.1
NM_001324331.1 Intron NP_001311260.1
NM_001324332.1 Intron NP_001311261.1
NM_001324333.1 Intron NP_001311262.1
NM_001324334.1 Intron NP_001311263.1
NM_002578.4 Intron NP_002569.1
XM_005262132.2 Intron XP_005262189.1
XM_005262133.3 Intron XP_005262190.1
XM_006724654.2 Intron XP_006724717.1
XM_006724655.2 Intron XP_006724718.1
XM_011530962.1 Intron XP_011529264.1
XM_011530967.2 Intron XP_011529269.1
XM_011530968.2 Intron XP_011529270.1
XM_011530969.1 Intron XP_011529271.1
XM_011530971.2 Intron XP_011529273.1
XM_017029557.1 Intron XP_016885046.1
XM_017029558.1 Intron XP_016885047.1
XM_017029559.1 Intron XP_016885048.1
XM_017029560.1 Intron XP_016885049.1
XM_017029561.1 Intron XP_016885050.1
XM_017029562.1 Intron XP_016885051.1
XM_017029563.1 Intron XP_016885052.1

View Full Product Details