Product Details

SNP ID
rs3848596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33144855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGTTGATTTGAGGATCCGGTGGA[C/T]GGTTCTGTGGTTCGCGATTTTGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR88 PubMed Links

Gene Details

Gene
WDR88
Gene Name
WD repeat domain 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173479.3 471 Silent Mutation GAC,GAT D133D NP_775750.3
XM_011526449.2 471 Silent Mutation GAC,GAT D133D XP_011524751.1
XM_011526450.2 471 Silent Mutation GAC,GAT D133D XP_011524752.1
XM_017026282.1 471 Silent Mutation GAC,GAT D133D XP_016881771.1
XM_017026283.1 471 Silent Mutation GAC,GAT D133D XP_016881772.1

View Full Product Details