Product Details

SNP ID
rs6701047
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:160409190 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTCAAGGAACCAGTGTCCAGAGC[A/G]GTCTCATGGAGAAACCTGGTCTCAC
Phenotype
MIM: 600533
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VANGL2 PubMed Links

Gene Details

Gene
VANGL2
Gene Name
VANGL planar cell polarity protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020335.2 Intron NP_065068.1
XM_005245357.1 Intron XP_005245414.1
XM_011509804.1 Intron XP_011508106.1
XM_011509805.2 Intron XP_011508107.1

View Full Product Details