Product Details

SNP ID
rs10219650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101745707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCATTGTATTATGACATAAAATA[C/T]GGCTAGACGCCAAGGTTTAACCATA
Phenotype
MIM: 616747 MIM: 607840 MIM: 604759
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHPT1 PubMed Links

Gene Details

Gene
CHPT1
Gene Name
choline phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020244.2 5317 Intron NP_064629.2
XM_011538574.1 5317 Intron XP_011536876.1
XM_011538575.1 5317 Intron XP_011536877.1
Gene
GNPTAB
Gene Name
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024312.4 5317 UTR 3 NP_077288.2
XM_006719593.2 5317 Intron XP_006719656.1
XM_011538731.2 5317 UTR 3 XP_011537033.1
XM_017019961.1 5317 UTR 3 XP_016875450.1
XM_017019962.1 5317 UTR 3 XP_016875451.1
Gene
SYCP3
Gene Name
synaptonemal complex protein 3
There are no transcripts associated with this gene.

View Full Product Details