Product Details

SNP ID
rs9532740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41191672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCCAATTTTTTCCTTTGTAGAA[A/G]AAAACCAAAATAATTTTACAAACTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KBTBD7 PubMed Links

Gene Details

Gene
KBTBD7
Gene Name
kelch repeat and BTB domain containing 7
There are no transcripts associated with this gene.

Gene
LOC101929140
Gene Name
uncharacterized LOC101929140
There are no transcripts associated with this gene.

View Full Product Details