Product Details

SNP ID
hCV30461635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40448195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCGGGATGGCTGGGCCCACTCA[G/T]TTCATAGCCTAGGTGTCTCTCGAGG
Phenotype
MIM: 146733
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IGFBP4 PubMed Links

Gene Details

Gene
IGFBP4
Gene Name
insulin like growth factor binding protein 4
There are no transcripts associated with this gene.

View Full Product Details