Product Details

SNP ID
rs10494241
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:145943179 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTTTAAACTGTCGACAGAATTAG[T/C]TACTCAAAAGTCAGATTCTGGCCGG
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
LIX1L PubMed Links

Gene Details

Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153713.2 Intron NP_714924.1
XM_017000316.1 Intron XP_016855805.1

View Full Product Details