Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001077188.1 | 2736 | UTR 3 | NP_001070656.1 | ||
NM_147175.3 | 2736 | UTR 3 | NP_671704.3 | ||
XM_005262490.3 | 2736 | UTR 3 | XP_005262547.1 | ||
XM_005262491.2 | 2736 | UTR 3 | XP_005262548.1 | ||
XM_011531406.1 | 2736 | UTR 3 | XP_011529708.1 | ||
XM_011531407.2 | 2736 | Intron | XP_011529709.1 | ||
XM_011531408.2 | 2736 | Intron | XP_011529710.1 | ||
XM_017029944.1 | 2736 | UTR 3 | XP_016885433.1 | ||
XM_017029945.1 | 2736 | UTR 3 | XP_016885434.1 | ||
XM_017029946.1 | 2736 | UTR 3 | XP_016885435.1 |