Product Details

SNP ID
rs9535794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51933012 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGGAAAACACAATCAACAGCGC[A/G]CTTGGATTTTATAAAATAAAACCCT
Phenotype
MIM: 606882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP7B PubMed Links

Gene Details

Gene
ATP7B
Gene Name
ATPase copper transporting beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000053.3 5678 UTR 3 NP_000044.2
NM_001005918.2 5678 UTR 3 NP_001005918.1
NM_001243182.1 5678 UTR 3 NP_001230111.1
XM_005266423.2 5678 UTR 3 XP_005266480.1
XM_005266424.4 5678 UTR 3 XP_005266481.1
XM_005266427.2 5678 UTR 3 XP_005266484.1
XM_005266428.1 5678 UTR 3 XP_005266485.1
XM_005266430.4 5678 UTR 3 XP_005266487.1
XM_005266431.3 5678 UTR 3 XP_005266488.1
XM_005266432.2 5678 UTR 3 XP_005266489.1
XM_006719837.3 5678 UTR 3 XP_006719900.1
XM_006719838.1 5678 UTR 3 XP_006719901.1
XM_006719839.1 5678 UTR 3 XP_006719902.1
XM_011535117.2 5678 UTR 3 XP_011533419.1
XM_011535118.1 5678 UTR 3 XP_011533420.1
XM_011535119.1 5678 UTR 3 XP_011533421.1
XM_011535122.2 5678 UTR 3 XP_011533424.1
XM_017020627.1 5678 UTR 3 XP_016876116.1
XM_017020628.1 5678 UTR 3 XP_016876117.1

View Full Product Details