Product Details

SNP ID
rs10245199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151061395 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGAGGGGGCACCCCCTCCCTGC[A/G]GGGGCATCTTGGGCTCTCTCCAGAG
Phenotype
MIM: 611741 MIM: 123831 MIM: 109280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASIC3 PubMed Links

Gene Details

Gene
ASIC3
Gene Name
acid sensing ion channel subunit 3
There are no transcripts associated with this gene.

Gene
CDK5
Gene Name
cyclin dependent kinase 5
There are no transcripts associated with this gene.

Gene
SLC4A2
Gene Name
solute carrier family 4 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199692.1 Intron NP_001186621.1
NM_001199693.1 Intron NP_001186622.1
NM_001199694.1 Intron NP_001186623.1
NM_003040.3 Intron NP_003031.3
XM_006716094.3 Intron XP_006716157.1

View Full Product Details