Product Details

SNP ID
rs6703241
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:201195089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCGGGCACCAAATAGTCACTCCT[C/T]CTCCTAGTTGTGATTCCTCCTCCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFN1 PubMed Links

Gene Details

Gene
IGFN1
Gene Name
immunoglobulin-like and fibronectin type III domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164586.1 Intron NP_001158058.1
XM_005245580.3 Intron XP_005245637.1
XM_006711618.3 Intron XP_006711681.1
XM_011510120.2 Intron XP_011508422.1
XM_011510121.2 Intron XP_011508423.1
XM_017002787.1 Intron XP_016858276.1

View Full Product Details