Product Details

SNP ID
rs10164293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10321725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACACCCCAACTCCAGGGCACGGC[C/G]GATCCTGGGCAGACAGGCACAGGGT
Phenotype
MIM: 614585 MIM: 609950
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FDX1L PubMed Links

Gene Details

Gene
FDX1L
Gene Name
ferredoxin 1 like
There are no transcripts associated with this gene.

Gene
RAVER1
Gene Name
ribonucleoprotein, PTB binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133452.2 Intron NP_597709.2
XM_011527671.1 Intron XP_011525973.1

View Full Product Details