Product Details

SNP ID
rs6061100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32015677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTTTGATAGTTCTTGTTATCA[A/G]GCGTTCATGCATGAGAATCCTCCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCM2L PubMed Links

Gene Details

Gene
CCM2L
Gene Name
CCM2 like scaffolding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080625.3 Intron NP_542192.2
XM_006723706.3 Intron XP_006723769.1
XM_011528566.1 Intron XP_011526868.1
XM_011528567.2 Intron XP_011526869.1
XM_011528568.2 Intron XP_011526870.1
XM_011528569.1 Intron XP_011526871.1

View Full Product Details