Product Details

SNP ID
rs10425282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50027448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAAGACTCTTGGGCCCAAGGTCT[C/T]TCAGCCTGATAATTGTTTGGCTTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VRK3 PubMed Links

Gene Details

Gene
VRK3
Gene Name
vaccinia related kinase 3
There are no transcripts associated with this gene.

Gene
ZNF473
Gene Name
zinc finger protein 473
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017026574.1 Intron XP_016882063.1

View Full Product Details