Product Details

SNP ID
rs9314442
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:241226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGTTGGGGAAATAAATTCACTC[A/G]TTACCTATGTTATTGGCAGGAGTGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RPL23AP53 PubMed Links

Gene Details

Gene
RPL23AP53
Gene Name
ribosomal protein L23a pseudogene 53
There are no transcripts associated with this gene.

Gene
ZNF596
Gene Name
zinc finger protein 596
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042415.2 Intron NP_001035880.1
NM_001042416.2 Intron NP_001035881.1
NM_001287254.1 Intron NP_001274183.1
NM_001287255.1 Intron NP_001274184.1
NM_001287256.1 Intron NP_001274185.1
NM_001287399.1 Intron NP_001274328.1
NM_173539.3 Intron NP_775810.2
XM_011534744.1 Intron XP_011533046.1
XM_017013166.1 Intron XP_016868655.1

View Full Product Details