Product Details

SNP ID
rs3856281
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:16397788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTATCTGCTACCATCAGCCCCT[G/T]CCAGAGCCCATCTGCCCCGCCCAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPATA21 PubMed Links

Gene Details

Gene
SPATA21
Gene Name
spermatogenesis associated 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198546.1 3335 Intron NP_940948.1
XM_011541407.2 3335 UTR 3 XP_011539709.1
XM_011541417.2 3335 Intron XP_011539719.1
XM_017001216.1 3335 Intron XP_016856705.1
XM_017001217.1 3335 Intron XP_016856706.1
XM_017001218.1 3335 Intron XP_016856707.1
XM_017001219.1 3335 Intron XP_016856708.1
XM_017001220.1 3335 Intron XP_016856709.1
XM_017001221.1 3335 Intron XP_016856710.1
XM_017001222.1 3335 Intron XP_016856711.1
XM_017001223.1 3335 Intron XP_016856712.1
XM_017001224.1 3335 UTR 3 XP_016856713.1
XM_017001225.1 3335 Intron XP_016856714.1
Gene
SZRD1
Gene Name
SUZ RNA binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114600.2 3335 UTR 3 NP_001108072.1
NM_001271869.1 3335 UTR 3 NP_001258798.1

View Full Product Details