Product Details

SNP ID
rs10425021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9447727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATGGGATGATGTAAAGTGATCA[A/C]ATTTACAAATAACTGGCATTCCAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372269 PubMed Links

Gene Details

Gene
LOC105372269
Gene Name
endogenous retrovirus group K member 8 Gag polyprotein-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011528461.2 Intron XP_011526763.1
Gene
ZNF560
Gene Name
zinc finger protein 560
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152476.2 Intron NP_689689.2
XM_011527696.2 Intron XP_011525998.1
XM_011527697.2 Intron XP_011525999.1
XM_017026327.1 Intron XP_016881816.1
XM_017026328.1 Intron XP_016881817.1
XM_017026329.1 Intron XP_016881818.1
XM_017026330.1 Intron XP_016881819.1

View Full Product Details