Product Details

SNP ID
rs6060714
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35954004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCTAGAGCCGGCGGGGCCTGGT[A/G]TGGAGCGCGCTTCAGCTTCGGCCTG
Phenotype
MIM: 610335 MIM: 610416
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CNBD2 PubMed Links

Gene Details

Gene
CNBD2
Gene Name
cyclic nucleotide binding domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207076.2 515 Intron NP_001194005.1
NM_001304367.1 515 Intron NP_001291296.1
NM_080834.3 515 Intron NP_543024.2
XM_005260295.2 515 Intron XP_005260352.1
XM_011528590.2 515 Intron XP_011526892.1
XM_011528592.1 515 Intron XP_011526894.1
XM_011528593.2 515 Intron XP_011526895.1
XM_011528598.1 515 Intron XP_011526900.1
XM_017027683.1 515 Intron XP_016883172.1
Gene
PHF20
Gene Name
PHD finger protein 20
There are no transcripts associated with this gene.

Gene
SCAND1
Gene Name
SCAN domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016558.3 515 Missense Mutation ACA,ATA T94I NP_057642.1
NM_033630.2 515 Missense Mutation ACA,ATA T157I NP_361012.2

View Full Product Details