Product Details

SNP ID
rs9948674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74541305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTGATCGGATCCTCCCCTGAG[C/T]GTGCACTGTCTAGGGCAGCTGTCCT
Phenotype
MIM: 609064
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNDP1 PubMed Links

Gene Details

Gene
CNDP1
Gene Name
carnosine dipeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032649.5 Intron NP_116038.4

View Full Product Details