Product Details
- SNP ID
-
rs9298071
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:63015791 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTACTCCCCCTGCAGCATATCCC[A/C]TGCTATGCTCACTCCGCAACTCATT
- Phenotype
-
MIM: 601509
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GGH
PubMed Links
Gene Details
- Gene
- GGH
- Gene Name
- gamma-glutamyl hydrolase
View Full Product Details