Product Details

SNP ID
rs10257334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130397972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTCAGCCCTGACAAAGGACTTC[C/G]GAGTCCTCTTCACCTTGTGTGTCCA
Phenotype
MIM: 610523 MIM: 114850
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEP41 PubMed Links

Gene Details

Gene
CEP41
Gene Name
centrosomal protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257158.1 2246 UTR 3 NP_001244087.1
NM_001257159.1 2246 UTR 3 NP_001244088.1
NM_001257160.1 2246 Intron NP_001244089.1
NM_018718.2 2246 UTR 3 NP_061188.1
XM_011516709.2 2246 UTR 3 XP_011515011.1
XM_011516710.2 2246 UTR 3 XP_011515012.1
Gene
CPA1
Gene Name
carboxypeptidase A1
There are no transcripts associated with this gene.

View Full Product Details