Product Details

SNP ID
rs10141220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50420898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGACTGATAGGAAAAAAAAAAA[C/G]AAGTACTATTCCAAACATTTTAGTT
Phenotype
MIM: 604923
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAP4K5 PubMed Links

Gene Details

Gene
MAP4K5
Gene Name
mitogen-activated protein kinase kinase kinase kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006575.4 Intron NP_006566.2
NM_198794.2 Intron NP_942089.1
XM_006720013.3 Intron XP_006720076.1
XM_011536377.2 Intron XP_011534679.1
XM_011536378.2 Intron XP_011534680.1
XM_011536379.2 Intron XP_011534681.1
XM_011536380.2 Intron XP_011534682.1

View Full Product Details